Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ Intrinsically Safe Enduro Pro HAZ-LO Headlamp, Spot/Flood Beam

Catalog No. 19028339 Shop All Streamlight Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-028-339 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-028-339 Supplier Streamlight™ Supplier No. 61424
Add to Cart
Add to Cart

Low-profile industrial headlamp with Class I, Division 1 safety ratings for use in hazardous locations. Three lighting modes cover a variety of tasks: spot for a focused beam, flood for soft area illumination that won't tire your eyes, and a spot/flood combo for even more light output.

  • Low-profile industrial headlamp with Class I, Division 1 safety ratings for use in hazardous locations
  • Three lighting modes cover a variety of tasks: spot for a focused beam, flood for soft area illumination that won't tire your eyes, and a spot/flood combo for even more light output
  • 200 lumens and 95m beam
  • Class 1, Div. 1 rated
  • Durable thermoplastic construction
  • 45 degree tilting head, especially handy for reading
  • IPX7 water resistant
  • Strapless mounting to hard hats
  • Push button switch with mode memory

Specifications

Voltage 1.5 V
Weight (English) 3.5 oz.
Weight (Metric) 0.099 kg
Battery Run Time 5 Hrs
Battery Type Alkaline
Beam Distance 73 m
Candle Power 1320 cd
Certifications/Compliance Class I, II, Division 1, Groups A, B, C, D, E, F, G; Class III; Class 1, Zone 0, Group IIC, Exia; T-Code: T4(approved for use with Energizer E92, Duracell MN2400, Rayovac NO0.824, and Panasonic LR03PA). Permissible Flashlight, Approval No. 20-A190001-0.Tested for Intrinsic Safety in Methane-Air mixtures only.(approved for use with Energizer E92, Duracell MN2400 and Rayovac NO0.824), ATEX: II 1 G Ex ia op is IIC T4 Ga †FTZU 18 ATEX 0101 XIECEx: Ex ia op is IIC T4 Ga - IECEx FTZU 19.0003 X, Meets applicable European Community Directives. Inmetro: Ex ia op is IIC T4 Ga - DNV 20.0025 X
Color Yellow
Description Intrinsically Safe
Includes (3) in.AAA in. Alkaline Batteries, Elastric headstrap, Rubber hard hat strap, 3 M Dual Lock fasteners, Extension Pole to 84 in.
Length (English) 1.5 in.
Lumen 235 lm (High), 160 lm (Low)
Material Thermoplastic
Warranty 2 year warranty with proof of purchase
Product Type Headlamp
Show More Show Less

Class I, II, Division 1, Groups A, B, C, D, E, F, G; Class III; Class 1, Zone 0, Group IIC, Exia; T-Code: T4(approved for use with Energizer E92, Duracell MN2400, Rayovac NO.824, and Panasonic LR03PA). Permissible Flashlight, Approval No. 20-A190001-0.Tested for Intrinsic Safety in Methane-Air mixtures only.(approved for use with Energizer E92, Duracell MN2400 and Rayovac NO.824), ATEX: II 1G Ex ia op is IIC T4 Ga FTZU 18 ATEX 0101XIECEx: Ex ia op is IIC T4 Ga - IECEx FTZU 19.0003X, Meets applicable European Community Directives. Inmetro: Ex ia op is IIC T4 Ga - DNV 20.0025 X

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.