Learn More
Cayman Chemical InTRleukIn12R2 RabbIt Mon 50uL
SDP
Catalog No. 502917998
Encompass_Preferred
A positive allosteric modulator of AMPA receptors potentiates glutamate-induced currents in HEK293 cells expressing human GluR2 homomeric AMPA receptors (EC50 2.51 UM) as well as HEK293 cells expressing GluR1 GluR3 or GluR4 homomeric AMPA receptors increases novel object exploration in a novel object recognition test in rats at 0.3 and 1 mg/kg reduces escape latencies in the Morris water maze in aged rats
Catalog No. 50-291-7998 Supplier Cayman Chemical Supplier No. 3809150 UL
A positive allosteric modulator of AMPA receptors potentiates glutamate-induced currents in HEK293 cells expressing human GluR2 homomeric AMPA receptors (EC50 2.51 UM) as well as HEK293 cells expressing GluR1 GluR3 or GluR4 homomeric AMPA receptors increases novel object exploration in a novel object recognition test in rats at 0.3 and 1 mg/kg reduces escape latencies in the Morris water maze in aged rats