Learn More
Cayman Chemical InTRleukIn16 MoncLNal nt 50uL
SDP
Catalog No. 502950759
Encompass_Preferred
An internal standard for the quantification of ochratoxin A by GC- or LC-MS
Catalog No. 50-295-0759 Supplier Cayman Chemical Supplier No. 3394950 UL
An internal standard for the quantification of ochratoxin A by GC- or LC-MS