Learn More
Cayman Chemical InTRleukIn6 MoncLNal ntI 200uL
SDP
Catalog No. 502947483
Encompass_Preferred
A mixture of bioactive sphingolipids found in mammalian cell membranes make up 2-15 of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells undergo hydrolysis by sphingomyelinase to form ceramides
Catalog No. 50-294-7483 Supplier Cayman Chemical Supplier No. 33947200 UL
A mixture of bioactive sphingolipids found in mammalian cell membranes make up 2-15 of the total organ phospholipid population but are found at higher concentrations in the brain and myelin sheaths surrounding peripheral nerves interact with cholesterol to control its distribution within cellular membranes and maintain cholesterol homeostasis in cells undergo hydrolysis by sphingomyelinase to form ceramides