Learn More
Cayman Chemical InTRleukIn6 RabbIt MoncLN 50uL
SDP
Catalog No. 502915242
Encompass_Preferred
A p70S6K1 inhibitor (IC50 52 nM) selective for p70S6K1 over a panel of 43 kinases at 1 UM
Catalog No. 50-291-5242 Supplier Cayman Chemical Supplier No. 3706850 UL
A p70S6K1 inhibitor (IC50 52 nM) selective for p70S6K1 over a panel of 43 kinases at 1 UM