Learn More
Med Vet International Dynarex Endotracheal Tube introducer (Bougie), Pediatric - 10 Fr, 10/Box
Catalog No. 502831597
Encompass_Preferred
Dynarex Resp-O2™ Endotracheal Tube Introducers, also known as Bougies, are safe and effective adjuncts to help facilitate difficult endotracheal intubations, including blind intubations. The sturdy Introducer is designed to slide into the endotracheal tube and guide it into the patient’s airway. The Introducer is constructed from high-quality plastic materials for optimal reliability and is clearly marked with calibrations to indicate the depth of the endotracheal tube for improved safety.  Dynarex Resp-O2™ Endotracheal Tube Introducers (Bougies) are premium medical devices designed to effectively guide and support endotracheal tubes during difficult intubation procedures. The sterile Introducer is disposable and comes individually wrapped for optimal hygiene.
Catalog No. 50-283-1597 Supplier Med Vet International Supplier No. 36260
Dynarex Resp-O2™ Endotracheal Tube Introducers, also known as Bougies, are safe and effective adjuncts to help facilitate difficult endotracheal intubations, including blind intubations. The sturdy Introducer is designed to slide into the endotracheal tube and guide it into the patient’s airway. The Introducer is constructed from high-quality plastic materials for optimal reliability and is clearly marked with calibrations to indicate the depth of the endotracheal tube for improved safety.  Dynarex Resp-O2™ Endotracheal Tube Introducers (Bougies) are premium medical devices designed to effectively guide and support endotracheal tubes during difficult intubation procedures. The sterile Introducer is disposable and comes individually wrapped for optimal hygiene.