Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Introduction to Basic, Selective, and Differential Media Kit
GREENER_CHOICESCI_ED

Catalog No. S18902 Shop All Aldon Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S18902 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S18902 Supplier Innovating Science™ Supplier No. IS3610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With the Introduction to Basic, Selective, and Differential Media Kit, students will learn about the different types of media that can be used to grow bacteria. They will make predictions, streak the different media with bacteria cultures, and evaluate bacteria growth.

  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: LS1.A, LS1.B
  • Performance Expectations: MS-LS1-1, MS-LS1-5
  • Cross Cutting Concepts: Cause and Effect, Scale, Proportion, and Quantity
  • Engineering Practices: Constructing Explanations and Designing Solutions, Planning and Carrying Out Investigations

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
Includes 200 mL Prepared Tryptic Soy Agar (2), 200 mL Prepared MacConkey Agar (2), Lyophilized Cultures of Serratia marcescens (10), Lyophilized Cultures of Bacillus subtilis (10), Lyophilized Cultures of Escherichia coli (10), Tryptic Soy Broth Tubes (30), Sterile Petri Dishes (40), Sterile Pipettors (30), Cotton Swab 2-Packs (40)
Class Size 10 Lab Groups
Product Type Life Science Microbiology Education Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.