Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Introduction to Chromatography Lab
SCI_ED

Catalog No. S67396
Change view
Click to view available options
40-student kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S67396 40-student kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S67396 Supplier Neo/Sci Corporation Supplier No. 201683

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Separate plant pigments and dye samples to learn about chromatography in clinical, forensic and analytical laboratories.

  • See the use of chromatography in identifying organic and inorganic compounds
  • Learn how solvent and substrate variables affect chromatography
  • Calculate the Rf values of unknown dye samples

Order Info

Class size: 40 students.

Specifications

For Use With (Application) Separates Plant Pigments and Dye Samples, Employed to Identify Organic and Inorganic Compounds
Class Size 40 Students
Product Type Lab Investigation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.