Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Introduction to Complexometric Titration
SCI_ED

Catalog No. S01993 Shop All Aldon Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01993 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING! : This product can expose you to chemicals including Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.
Catalog No. S01993 Supplier Innovating Science™ Supplier No. IS2534
Add to Cart
Add to Cart
WARNING! : This product can expose you to chemicals including Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.

Introduce topics such as metal-ligand interactions in this activity using complexometric titration to determine the calcium content of both known and unknown samples.

Complex formation is frequently used in both qualitative quantitative inorganic analysis. Introduce topics such as metal-ligand interactions in this activity using complexometric titration to determine the calcium content of both known and unknown samples.

Specifications

Grade 9 to Undergraduate
Class Size 15 Lab Groups
Product Type Structure and Concentration In Acid and Base Titrations Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.