Learn More
Introduction to Fossils Poster
SCI_ED
Catalog No. S01958L Shop All Fischer Technical Company Products
Change view
Click to view available options
Laminated; Legend: Introduction to Fossils
Catalog No.
S01958L Laminated; Legend: Introduction to Fossils
1 options
Catalog No. S01958L Supplier Fischer Technical Company Supplier No. E103L
Explains what fossils are and how a few pioneering scientists eventually realized that they were the remains or traces of once-living organisms.
  • Fossil expeditions discuss what fossils tell us
  • Relationship of fossils to geology
  • Covers the principle of original horizontality, the law of superimposition, the principle of faunal succession, geological time charts, index fossils and both relative and absolute dating
  • Size: 24 x 36 in. (60.96 x 91.44cm)
  • Laminated
Specifications
Format Poster
Product Type Introduction to Fossils Poster
For Use With (Application) Covers the principle of original horizontality, the law of superimposition, the principle of faunal succession, geological time charts, index fossils and both relative and absolute dating
Subject Social Science