Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Introduction to Materials Science
GREENER_CHOICESCI_ED

Catalog No. S280004 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S280004 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S280004 Supplier Innovating Science™ Supplier No. IS3470
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Introduction to Materials Science introduces students to the field of materials science as they explore the four main types of materials: metals, polymers, ceramics, and composites.

  • In this activity, students will be introduced to the field of materials science as they explore the four main types of materials: metals, polymers, ceramics, and composites.
  • They will test the physical and mechanical properties of different material samples, then categorize each material based on their results.
  • Kit contains enough material for 15 groups.
  • Teacher's Guide and Student Study Guide copymasters included.

Specifications

Certifications/Compliance Next Generation Science Standards (NGSS)
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Materials Science
Includes 30 Aluminum Strips, 30 Copper Strips, 30 Tongue Depressors, 30 Playing Cards, 30 Tile Pieces, 30 Glass Slides, 30 Plastic Chips, 30 Balloons, 30 Plastic Bags, 15 Steel Nails
Class Size 15 Lab Groups
Product Type Mini Classroom Curriculum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.