Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Introduction to Streak Isolation and Aseptic Technique Kit
GREENER_CHOICESCI_ED

Catalog No. S18903 Shop All Aldon Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S18903 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S18903 Supplier Innovating Science™ Supplier No. IS3611
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With the Introduction to Streak Isolation and Aseptic Technique Kit, students will be introduced to the skills and techniques that microbiologists use on a daily basis. Students will perform a streak isolation with the provided bacterial cultures, as well as samples they collect.

  • Students will practice performing a streak isolation with the provided bacterial cultures, as well as samples they collect from their classroom, to learn about different types of media and how bacterium reproduce
  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: LS1.A
  • Performance Expectations: MS-LS1-1
  • Cross Cutting Concepts: Scale, Proportion, and Quantity
  • Engineering Practices: Planning and Carrying Out Investigations

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
Includes 200 mL Prepared Tryptic Soy Agar (2), Lyophilized Cultures of Serratia marcescens (10), Sterile Petri Dishes (20), Tryptic Soy Broth Tubes (10), Inoculation Loops (10), Sterile Pipettors (10), Sterile Cotton Swab 2-Packs (10)
Class Size 10 Lab Groups
Product Type Life Science Microbiology Education Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.