Learn More
Introductory Fossil Collection
SDPSCI_ED
Catalog No. S45057
Click to view available options
Product Type:
Introductory Fossil Collection
Catalog No. Product Type
S45057 Introductory Fossil Collection
1 options
Catalog No. S45057 Supplier Geoscience Industries Supplier No. GEO FOS 20
20 fossils offer broad array of geological time.
With some samples ranging from more than 400 million years old, this collection is displayed in a durable, compartmented box.
  • Illustrated key gives pertinent facts about each fossil including its geological time
  • 20 selected fossils including foraminifer, horn coral, snails, trilobite, shark's tooth, bryozoan, brachiopod, cephalopod, clams, and oyster
Specifications
For Use With (Application) 20 Fossils offer broad array of geological time
Includes Foraminifer, horn coral, snails, trilobite, shark's tooth, bryozoan, brachiopod, cephalopod, clams, oyster; Includes illustrated key
Product Type Introductory Fossil Collection