Learn More
Maine Molecular Quality Controls Inc INTROL CF Panel II Control
SDP
Catalog No. NC3886252
Encompass
INTROL CF Panel II Control is intended for in vitro use as a quality control to monitor analytical performance of the extraction, amplification and detection steps of test systems used in the qualitative measurement of the Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) gene. This product is intended to be extracted and analyzed routinely with each cystic fibrosis (CF) test run. This product is designed to monitor the detection of 54 CFTR mutations, genetic risk factors associated with cystic fibrosis, including the original 23 mutations recommended for testing by ACMG and ACOG. INTROL CF Panel II Control also monitors variants I506V, I507V, F508C, 5/7/9/11T and I148T. G110 contains: 3 bottles, 2 milliliters each.
Catalog No. NC3886252 Supplier Maine Molecular Quality Controls Inc Supplier No. G110
INTROL CF Panel II Control is intended for in vitro use as a quality control to monitor analytical performance of the extraction, amplification and detection steps of test systems used in the qualitative measurement of the Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) gene. This product is intended to be extracted and analyzed routinely with each cystic fibrosis (CF) test run. This product is designed to monitor the detection of 54 CFTR mutations, genetic risk factors associated with cystic fibrosis, including the original 23 mutations recommended for testing by ACMG and ACOG. INTROL CF Panel II Control also monitors variants I506V, I507V, F508C, 5/7/9/11T and I148T. G110 contains: 3 bottles, 2 milliliters each.