Learn More
Maine Molecular Quality Controls Inc INTROL TB Panel M114
SDP
Catalog No. NC0574885
Encompass
INTROL TB Panel M114 is intended for use as a quality control to monitor analytical performance of M. tuberculosis (MTB) on the GeneXpert System. INTROL TB Panel M114 is formulated to monitor the detection of MTB and multi-drug resistant MTB (MDR-TB) mutations. INTROL TB Panel M114 (P/N: M114-5) is comprised of 5 kits; each kit is comprised of three bottles, 1 mL each. Two bottles contain encapsulated MTB DNA, gene segments IS6110, hsp65, 16S rRNA, 23S rRNA, inhA, katG, and rpoB. The third bottle contains buffer and preservative only.
Catalog No. NC0574885 Supplier Maine Molecular Quality Controls Inc Supplier No. M1145
INTROL TB Panel M114 is intended for use as a quality control to monitor analytical performance of M. tuberculosis (MTB) on the GeneXpert System. INTROL TB Panel M114 is formulated to monitor the detection of MTB and multi-drug resistant MTB (MDR-TB) mutations. INTROL TB Panel M114 (P/N: M114-5) is comprised of 5 kits; each kit is comprised of three bottles, 1 mL each. Two bottles contain encapsulated MTB DNA, gene segments IS6110, hsp65, 16S rRNA, 23S rRNA, inhA, katG, and rpoB. The third bottle contains buffer and preservative only.