Learn More
Maine Molecular Quality Controls Inc INTROL TB Panel M114
SDP
Supplier:  Maine Molecular Quality Controls Inc M11410
Encompass
INTROL TB Panel M114 is intended for use as a quality control to monitor analytical performance of M. tuberculosis (MTB) on the GeneXpert System. INTROL TB Panel M114 is formulated to monitor the detection of MTB and multi-drug resistant MTB (MDR-TB) mutations. INTROL TB Panel M114 (P/N: M114-10) is comprised of 10 kits; each kit is comprised of three bottles, 1 mL each. Two bottles contain encapsulated MTB DNA, gene segments IS6110, hsp65, 16S rRNA, 23S rRNA, inhA, katG, and rpoB. The third bottle contains buffer and preservative only.
Catalog No. NC0565040