Learn More
Pyramex™ Intruder™ Readers Safety Eyewear
Catalog No. 19155563 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Magnification Power:
+1.50
+2.00
+2.50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-155-563 +1.50
19-155-564 +2.00
19-155-565 +2.50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-563 Supplier Pyramex™ Supplier No. S4110R15
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Integrated nosepiece, lightweight, frameless protection
  • Lens insert is out of user's direct line of vision
  • Lightweight, frameless protection
  • Superior comfort and fit
  • Mini Intruder has all the great features of Intruder in a design better suited for smaller facial size
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Clear
    Frame Color Clear
    Magnification Power +1.50