Learn More
Fine Science Tools Instrument Tip Protectors, Blue, 2 x 19 mm
SDP
Catalog No. NC9190335
Encompass
Product: Instrument Tip Protectors - 2 x 19mm/Blue/ Dimensions: 2 x 19mm/ Color: Blue/ Material: Plastisol Autoclave Safe: Yes/ Unit: 25
Catalog No. NC9190335 Supplier Fine Science Tools Supplier No. 2906002
Product: Instrument Tip Protectors - 2 x 19mm/Blue/ Dimensions: 2 x 19mm/ Color: Blue/ Material: Plastisol Autoclave Safe: Yes/ Unit: 25