Learn More
Aviva Systems Biology Corp INTS12 Antibody - N-terminal region (ARP51223_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51223_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INTS12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human INTS12. Alternate names: INT12, PHF22, SBBI22ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60520