Learn More
Aviva Systems Biology Corp INTS4 Antibody - middle region (ARP51694_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51694_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INTS4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human INTS4. Alternate names: INT4, MGC16733, MST093ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60588