Learn More
Peak Scientific Instruments Ltd Intura H2 1500 GC Carrier Gas Produces up to 1500cc/min of H2
Supplier: Peak Scientific Instruments Ltd 3305698005
99.99999% hydrogenVariable flow up to 1500cc/minVariable pressure up to 175psiLatest PEM technology, generating reliable and consistent source of hydrogenCompact design, saving valuable lab bench spaceDeionized water quality monitoring and alertsSafe gas supply with low internal H2 volume, internal leak detection and auto-shutdownTouchscreen for operation, reminders and alertsPressure water-fill option for additional ease of useLow power consumption and standby modeRemote start/stop functionality available for user configurationReady on-board daisy-chain (cascading) function for capacity expansionH2 detector compatibility as standard12-month comprehensive warranty, with the option to extend up to 5 years3-year PEM cell warranty as standard