Learn More
Peak Scientific Instruments Ltd Intura H2 500 GC Carrier Gas, Pressurized Produces up to 500cc/min of H2
SDP
Catalog No. 5000804202
Encompass_Preferred
99.99999% hydrogen
Variable flow up to 500cc/min
Variable pressure up to 175psi
Latest PEM technology, generating reliable and consistent source of hydrogen
Compact design, saving valuable lab bench space
Deionized water quality monitoring and alerts
Safe gas supply with low internal H2 volume, internal leak detection and auto-shutdown
Touchscreen for operation, reminders and alerts
Pressure water-fill option for additional ease of use
Low power consumption and standby mode
Remote start/stop functionality available for user configuration
Ready on-board daisy-chain (cascading) function for capacity expansion
H2 detector compatibility as standard
12-month comprehensive warranty, with the option to extend up to 5 years
3-year PEM cell warranty as standard
Catalog No. 50-008-04202 Supplier Peak Scientific Instruments Ltd Supplier No. 3305698013
99.99999% hydrogen
Variable flow up to 500cc/min
Variable pressure up to 175psi
Latest PEM technology, generating reliable and consistent source of hydrogen
Compact design, saving valuable lab bench space
Deionized water quality monitoring and alerts
Safe gas supply with low internal H2 volume, internal leak detection and auto-shutdown
Touchscreen for operation, reminders and alerts
Pressure water-fill option for additional ease of use
Low power consumption and standby mode
Remote start/stop functionality available for user configuration
Ready on-board daisy-chain (cascading) function for capacity expansion
H2 detector compatibility as standard
12-month comprehensive warranty, with the option to extend up to 5 years
3-year PEM cell warranty as standard