Learn More
Peak Scientific Instruments Ltd Intura N2 600 GC Gas Generator Produces up to 600cc/min of N2
SDP
Catalog No. 5000804186
Encompass_Preferred
Up to 600cc/min UHP N2 at up to 100psi
Up to >99.9995% nitrogen purity
<10ppm CO2
Purity safeguard, ensuring UHP gas supply
High-efficiency PSA purifier
Low power consumption, reducing power draw and operational carbon footprint
Compact design, saving valuable lab bench space
Ultra-low power consumption
12-month comprehensive warranty, with the option to extend up to 5 years
Catalog No. 50-008-04186 Supplier Peak Scientific Instruments Ltd Supplier No. 3305330002
Up to 600cc/min UHP N2 at up to 100psi
Up to >99.9995% nitrogen purity
<10ppm CO2
Purity safeguard, ensuring UHP gas supply
High-efficiency PSA purifier
Low power consumption, reducing power draw and operational carbon footprint
Compact design, saving valuable lab bench space
Ultra-low power consumption
12-month comprehensive warranty, with the option to extend up to 5 years