Learn More
Peak Scientific Instruments Ltd Intura N2|ZA 250 GC Gas Generator Produces up to 250cc/min N2 and 1500cc/min Air
SDP
Catalog No. 5000804209
Encompass_Preferred
Smallest 2-in-1 Nitrogen & Zero Air generator with Integrated air compressor
Up to 250cc/min UHP N2 and 1.5LPM Zero air
Ultra-fast start-up - High purity Nitrogen produced in under 45 mins
Patented AirMax™ intake filter
Purity safeguard feature
Compact design, saving valuable lab bench space
Improved, more efficient CAT chamber
Low power consumption, reducing power draw and operational carbon footprint
Highest pressure output from compressor-based UHP N2 & ZA generator
High-quality touchscreen for operation, reminders and alerts
12-month comprehensive warranty, with the option to extend up to 5 years
Catalog No. 50-008-04209 Supplier Peak Scientific Instruments Ltd Supplier No. 3305329101
Smallest 2-in-1 Nitrogen & Zero Air generator with Integrated air compressor
Up to 250cc/min UHP N2 and 1.5LPM Zero air
Ultra-fast start-up - High purity Nitrogen produced in under 45 mins
Patented AirMax™ intake filter
Purity safeguard feature
Compact design, saving valuable lab bench space
Improved, more efficient CAT chamber
Low power consumption, reducing power draw and operational carbon footprint
Highest pressure output from compressor-based UHP N2 & ZA generator
High-quality touchscreen for operation, reminders and alerts
12-month comprehensive warranty, with the option to extend up to 5 years