Learn More
Medchemexpress LLC HY-N0896 5mg Medchemexpress, Inulicin CAS:33627-41-7 Purity:>98%
SDP
Catalog No. 501933781
Encompass_Preferred
Medchemexpress, HY-N0896 5mg Inulicin CAS:33627-41-7 Inulicin (1-O-Acetylbritannilactone) is an active compound isolated from Inula Britannica L. Inulicin (1-O-Acetylbritannilactone) inhibits VEGF-mediated activation of Src and FAK . Inulicin (1-O-Acetylbritannilactone) inhibits LPS-induced PGE 2 production and COX-2 expression, and NF-κB activation and translocation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3781 Supplier Medchemexpress LLC Supplier No. HYN08965MG
Medchemexpress, HY-N0896 5mg Inulicin CAS:33627-41-7 Inulicin (1-O-Acetylbritannilactone) is an active compound isolated from Inula Britannica L. Inulicin (1-O-Acetylbritannilactone) inhibits VEGF-mediated activation of Src and FAK . Inulicin (1-O-Acetylbritannilactone) inhibits LPS-induced PGE 2 production and COX-2 expression, and NF-κB activation and translocation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.