Learn More
Biophysics Assay Laboratory Inc BioPAL Inulin FIT-GFR™ Kit, Accurate GFR Measurement, Research Use Only, 96-Well Test Kit
SDP
Catalog No. NC3359868
Encompass
The BioPAL INULIN FIT-GFR™ Kit offers a reliable solution for accurately measuring glomerular filtration rate (GFR) using inulin, recognized as the gold standard filtration marker. This enzyme immunoassay provides a sensitive method that overcomes the limitations of other techniques, making it indispensable for research in various fields such as physiology and pharmacology, with the capacity to support up to 15 GFR measurements in a 96-well format.
Catalog No. NC3359868 Supplier Biophysics Assay Laboratory Inc Supplier No. FIT0415
The BioPAL INULIN FIT-GFR™ Kit offers a reliable solution for accurately measuring glomerular filtration rate (GFR) using inulin, recognized as the gold standard filtration marker. This enzyme immunoassay provides a sensitive method that overcomes the limitations of other techniques, making it indispensable for research in various fields such as physiology and pharmacology, with the capacity to support up to 15 GFR measurements in a 96-well format.