Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inulin, MP Biomedicals™

Catalog No. ICN19897125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19897125 25 g
ICN19897180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19897125 Supplier MP Biomedicals, Inc Supplier No. 0219897125
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9005-80-5
Molecular Formula (C6H10O5)n
Synonym Dahlin

Specifications

Color White
Quantity 25 g
Formula Weight ≈5,000
Physical Form Free-flowing Powder
Chemical Name or Material Inulin
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.