Learn More
Medchemexpress LLC Inupadenant hydrochloride | 2411004-22-1 | 99.6% | 641.11 | 50 MG
SDP
Catalog No. 5000350405
Encompass_Preferred
Inupadenant hydrochloride (EOS-850) is an orally active, highly selective A2A receptor antagonist. It cannot cross the blood-brain barrier (BBB) and can enhance the humoral immune response, exhibiting anti-tumor activity.
  • Increases OVA-specific IgGs in the blood and tumors
  • Increases B cells and antibody-secreting cells in the spleen and tumors of CT26-OVA mice
Catalog No. 50-003-50405 Supplier Medchemexpress LLC Supplier No. HY137442A50MG
Inupadenant hydrochloride (EOS-850) is an orally active, highly selective A2A receptor antagonist. It cannot cross the blood-brain barrier (BBB) and can enhance the humoral immune response, exhibiting anti-tumor activity.
  • Increases OVA-specific IgGs in the blood and tumors
  • Increases B cells and antibody-secreting cells in the spleen and tumors of CT26-OVA mice