Learn More
Medchemexpress LLC Inupadenant hydrochloride | 2411004-22-1 | 99.6% | 641.11 g·mol⁻¹ | C25H27ClF2N8O4S2 | 5 MG
SDP
Catalog No. 5000221895
Encompass_Preferred
Inupadenant hydrochloride is the hydrochloride salt of inupadenant (EOS-850), an orally active, highly selective adenosine A2A receptor antagonist reported to be non-brain-penetrant and to enhance humoral immune responses with anti-tumor activity in preclinical studies. Provided as a purified research compound for pharmacology and immuno-oncology investigations.
  • Selective A2A receptor antagonist
  • Orally active compound
  • Does not cross the blood-brain barrier
  • High purity (99.6%)
  • Supplied in small research quantities suitable for in vivo and in vitro studies
Catalog No. 50-002-21895 Supplier Medchemexpress LLC Supplier No. HY137442A5MG
Inupadenant hydrochloride is the hydrochloride salt of inupadenant (EOS-850), an orally active, highly selective adenosine A2A receptor antagonist reported to be non-brain-penetrant and to enhance humoral immune responses with anti-tumor activity in preclinical studies. Provided as a purified research compound for pharmacology and immuno-oncology investigations.
  • Selective A2A receptor antagonist
  • Orally active compound
  • Does not cross the blood-brain barrier
  • High purity (99.6%)
  • Supplied in small research quantities suitable for in vivo and in vitro studies