Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Tape Logic Label, Fluorescent Red, 1x1 in, 500/Roll

Catalog No. 501917721
Encompass

This product consists of bright fluorescent red inventory labels made from paper, designed to quickly and easily identify shipments, pallets, shelves, or boxes. The labels feature a pressure-sensitive permanent adhesive and are easy to write on with pens or markers.

  • Type: Inventory labels
  • Color: Fluorescent red
  • Material: Paper
  • Width: 1 in
  • Length: 1 in
  • Number of labels per roll: 500
  • Quantity per roll: 1 roll
  • For use with: Multi-use

Catalog No. 50-191-7721 Supplier MSC Supplier No. 39572961
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product consists of bright fluorescent red inventory labels made from paper, designed to quickly and easily identify shipments, pallets, shelves, or boxes. The labels feature a pressure-sensitive permanent adhesive and are easy to write on with pens or markers.

  • Type: Inventory labels
  • Color: Fluorescent red
  • Material: Paper
  • Width: 1 in
  • Length: 1 in
  • Number of labels per roll: 500
  • Quantity per roll: 1 roll
  • For use with: Multi-use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.