Learn More
MSC Rockwell Labs Ltd. Bio Sanitation Foam, Hot Spot+, 19 oz, Aerosol
Supplier:  MSC 74333899
Encompass
This super-concentrated foaming formula, delivered in a 19 oz aerosol, contains a blend of premium, natural microbes and citrus oil. It is designed to eliminate organic build-up and odors in commercial facilities, effectively working as a green cleaner. The powerful probiotics in the foam activate upon contact with organic debris, actively digesting organic matter.
  • Eliminates organic build-up and odors
  • Contains natural microbes and citrus oil
  • Probiotics digest organic matter
  • Foam provides longer contact time
  • Suitable for treating drains, baseboards, and other areas with organic debris build-up
  • Applicable for food and non-food areas
  • Free of harsh chemicals or fumes
  • Does not harm septic systems or municipal water treatment operations
  • Green Zone™ product, suitable for green service programs
Catalog No. 50-283-9286