Learn More
Sigma Aldrich Invar™ - Controlled Expansion Alloy, Fe64Ni36
Catalog No. 503117951 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
10 Ea.
5 Ea.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-311-7951 10 Ea.
50-311-7952 1 Ea.
50-311-7966 5 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-311-7951 Supplier Sigma Aldrich Supplier No. GF0582671910EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Invar™ - Controlled Expansion Alloy, Fe64Ni36
Specifications
Quantity 10 Ea.
Synonym FE020265