Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ Inventus Biotech Post-Mortem Waxed Linen Suture Thread
SDP

Catalog No. 21520139
Change view
Click to view available options
Quantity:
1 lb. spool
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-139 1 lb. spool
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-139 Supplier Cenmed™ Supplier No. 8799901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enhanced nylon thread for strength and quality

  • Smooth 7 cord waxed thread
  • 1 pound spool
  • Excellent tensile strength and uniform lay
  • High quality suture cord

Specifications

Material Linen
Product Type Linen Suture Thread
Quantity 1 lb. spool
For Use With (Application) Post Mortem
Color Natural
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.