Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alere Inverness Clearview™ Mono-plus II Test

Catalog No. WA6908KCV Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
30 Cassettes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WA-6908KCV 30 Cassettes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WA-6908KCV Supplier Alere Supplier No. 6908KCV

missing translation for 'validMainframeMsg'

Includes

30 individually pouched test devices each containing one disposable transfer pipette, a developer buffer, positive and negative controls, two directional inserts (one for waived and one for non-waived) and one procedure card

Patented technology provides highly sensitive and specific results for the detection of infectious mononucleos

CLIA Complexity: Waived for whole blood; Moderate for serum or plasma

  • Sensitivity: 98.6%
  • Specificity: 98.8%
  • Simple two-step procedure
  • Built-in control ensures accuracy
  • Use with multiple sample tyes - whole blood, serum, or plasma
  • Positive results in as soon as 3 minutes; negative results, less than 5 minutes
  • Room temperature storage with 18 months shelf life

Specifications

Quantity 30 Cassettes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.