Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LACTOFERRIN CHEK™

Catalog No. 23046936
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-936 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-936 Supplier Techlab Inc Supplier No. 30352
Add to Cart
Add to Cart
This item is not returnable. View return policy

For qualitative detection of elevated levels of fecal lactoferrin, a marker of fecal leukocytes and an indicator of intestinal inflammation, to distinguish active IBD from IBS. Developed and Manufactured by TECHLAB, Inc.

  • 96-well microplate format eliminates need for manual smear
  • Assay can be run on the benchtop or automated with Dynex automated systems
  • Break-apart microplate wells minimize reagent use
  • Works with any specimen consistency, and samples can be refrigerated or stored at room temperature with no loss of sensitivity for two weeks

Specifications

Detectable Analytes Fecal Lactoferrin
Format Microplate
Type LACTOFERRIN CHEK™ Diagnostic Test Kit
Quantity 96 Tests
Test Time 75 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.