Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LACTOFERRIN SCAN™

Catalog No. 23046937
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-937 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-046-937 Supplier Techlab Inc Supplier No. 30351
Add to Cart
Add to Cart

The LACTOFERRIN-SCAN test is a quantitative ELISA for measuring concentrations of fecal lactoferrin, a marker for fecal leukocytes and an idicator of intestinal inflammation. The test can be used as an aid in assessing when an IBD patient is in remission and has responded to treatment. Developed and Manufactured by TECHLAB, Inc.

  • The test can be used as an in vitro diagnostic aid to distinguish patients with active inflammatory bowel disease (IBD) from those with noninflammatory irritable bowel syndrome (IBS)
  • The test can be used to assess when an IBD patient is in confirmed remission and has responded to treatment.
  • Standard curve allows for exact quantification of lactoferrin in samples
  • Assay can be run on the benchtop or automated with Dynex™ automated systems    
  • Works with any sample consistency
  • Samples can be refrigerated or stored at room temperature with no loss of sensitivity for two weeks
  • Yields results in 75 minutes
  • 96-well microplate format

Specifications

Detectable Analytes Fecal Lactoferrin
Format Microplate
Type LACTOFERRIN-SCAN™ Diagnostic Test Kit
Includes Stop solution, 20X Wash Buffer, conjugate, 10X Diluent, positive control, Lactoferrin standards (5), substrate, microassay plate
Quantity 96 Tests
Test Time 75 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.