Learn More
Chemglass Life Sciences Adapter, Vacuum/Inert Gas, 14/20 Inner Joint, PTFE Stpk, Airfree, Schlenk
SDP
Catalog No. AF0509B03
Encompass_Preferred
Adapter, Vacuum/Inert Gas, 14/20 Inner Joint, PTFE Stpk, Airfree, Schlenk. Similar to AF-0509, adapter has a T-Bore stopcock permitting vessels having a standard taper outer joint to be purged and evacuated. Available with a choice of a 4mm glass high vacuum stopcock having a Stainless Steel constant pressure retention device or 4mm PTFE stopcock.
Catalog No. AF0509B03 Supplier Chemglass Life Sciences Supplier No. AF0509B03
Adapter, Vacuum/Inert Gas, 14/20 Inner Joint, PTFE Stpk, Airfree, Schlenk. Similar to AF-0509, adapter has a T-Bore stopcock permitting vessels having a standard taper outer joint to be purged and evacuated. Available with a choice of a 4mm glass high vacuum stopcock having a Stainless Steel constant pressure retention device or 4mm PTFE stopcock.