Learn More
Medchemexpress LLC Invertase, baker's yeast (S. cerevisiae) | 9001-57-4 | ≥200U/mg solid | 270 kDa | 1 G
SDP
Catalog No. 5000400132
Encompass_Preferred
Invertase, baker's yeast (S. cerevisiae) is a major enzyme found in plants and microorganisms, often used in biochemical studies. It catalyzes the hydrolysis of the disaccharide sucrose into glucose and fructose. This product is obtained through deep microbial fermentation and is used to catalyze the hydrolysis of sucrose.
  • Effective temperature range 20-60°C
  • Optimal temperature 45-55°C
  • Effective pH range 3.0-8.0
  • Optimal pH 4.5-5.5
  • Inhibitors include Fe3+, Cu2+, Hg+, Pb+
  • Soluble in water
Catalog No. 50-004-00132 Supplier Medchemexpress LLC Supplier No. HYP29791G
Invertase, baker's yeast (S. cerevisiae) is a major enzyme found in plants and microorganisms, often used in biochemical studies. It catalyzes the hydrolysis of the disaccharide sucrose into glucose and fructose. This product is obtained through deep microbial fermentation and is used to catalyze the hydrolysis of sucrose.
  • Effective temperature range 20-60°C
  • Optimal temperature 45-55°C
  • Effective pH range 3.0-8.0
  • Optimal pH 4.5-5.5
  • Inhibitors include Fe3+, Cu2+, Hg+, Pb+
  • Soluble in water