Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Invertase, baker's yeast (S. cerevisiae) | 9001-57-4 | 270 kDa | 250 MG
SDP

Catalog No. 5000397294
Encompass_Preferred

Invertase, baker's yeast (S. cerevisiae) is a major enzyme found in plants and microorganisms, commonly used in biochemical studies. This enzyme catalyzes the hydrolysis of the disaccharide sucrose into glucose and fructose. Obtained through deep microbial fermentation, it is utilized to catalyze sucrose hydrolysis.

  • Effective temperature range: 20-60°C (optimal 45-55°C)
  • Effective pH range: 3.0-8.0 (optimal 4.5-5.5)
  • Soluble in water
  • Appearance: solid, white to off-white color

Catalog No. 50-003-97294 Supplier Medchemexpress LLC Supplier No. 250MGHYP2979
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Invertase, baker's yeast (S. cerevisiae) is a major enzyme found in plants and microorganisms, commonly used in biochemical studies. This enzyme catalyzes the hydrolysis of the disaccharide sucrose into glucose and fructose. Obtained through deep microbial fermentation, it is utilized to catalyze sucrose hydrolysis.

  • Effective temperature range: 20-60°C (optimal 45-55°C)
  • Effective pH range: 3.0-8.0 (optimal 4.5-5.5)
  • Soluble in water
  • Appearance: solid, white to off-white color

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.