Learn More
Chemglass Life Sciences Inverted Liquid Transfer Cap, for NEST 3L High Efficiency Erlenmeyer Flasks
SDP
Catalog No. 502733457
Encompass_Preferred
The inverted transfer cap is used along with a 2L, 3L or 5L shake flask to connect a liquid inlet tubing with the required device.  When liquid transfer is required, the liquid is transferred under gravity with the inverted Erlenmeyer flask.  Since inverted liquid transfer cap system do not require auxiliary device when transfer liquid. For some end-users who are worried about cell damage by using peristaltic pumps to transfer liquid, inverted liquid transfer cap system is one of their best choices. Closed transfer reduces the risks of contamination during liquid transfer. The cap is molded and connected in an integrated manner, reducing the risks of leakage and media residues. A variety of tubing diameters are available and aseptic welding of liquid inlet tubing under normal conditions is supported. High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance. Sterility level (SAL) is maintained at 10-6. Endotoxin-free, and no ingredients of ani
Catalog No. 50-273-3457 Supplier Chemglass Life Sciences Supplier No. CGN2165586
Add to Cart
Add to Cart
The inverted transfer cap is used along with a 2L, 3L or 5L shake flask to connect a liquid inlet tubing with the required device.  When liquid transfer is required, the liquid is transferred under gravity with the inverted Erlenmeyer flask.  Since inverted liquid transfer cap system do not require auxiliary device when transfer liquid. For some end-users who are worried about cell damage by using peristaltic pumps to transfer liquid, inverted liquid transfer cap system is one of their best choices. Closed transfer reduces the risks of contamination during liquid transfer. The cap is molded and connected in an integrated manner, reducing the risks of leakage and media residues. A variety of tubing diameters are available and aseptic welding of liquid inlet tubing under normal conditions is supported. High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance. Sterility level (SAL) is maintained at 10-6. Endotoxin-free, and no ingredients of ani