Learn More
ACCU-SCOPE™ Inverted Metallurgical Microscope
SDP
Catalog No. 01189257 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Eyepiece:
WF 10X/23 mm
Catalog No. Eyepiece
01-189-257 WF 10X/23 mm
1 options
Catalog No. 01-189-257 Supplier ACCU-SCOPE™ Supplier No. 14620
Add to Cart
Add to Cart
Request a Quote
The UNITRON™ 14620 inverted metallurgical microscope features a trinocular viewing head. Eyepiece accepts a 24 mm reticle infinity m plan achromat LWD 5x,10x, 20xr and semi-apo 50xr objectives, quintuple nosepiece, 5 watt variable led illuminator, coaxial coarse and fine focusing controls.
  • Trinocular viewing head, inclined 45° viewing split: 100% binocular, or 20% binocular / 80% camera
  • Widefield 10x/22 mm focusing eyepieces, eye cushions, eyepieces accept 26.5 mm reticles
  • 54 to 75 mm interpupillary adjustment
  • Infinity M plan achromat LWD 5x, 10x, 20xR, and semi-Apo 50x objectives
  • Quintuple revolving nosepiece with vertical action movement
  • Brightfield and simple polarizer/analyzer
  • Attachable mechanical stage, with right hand coaxial controls, travel range: 120 mm (X) 78mm (Y)
  • Built-in lever type condenser
  • Coaxial coarse and fine focusing controls
  • 5-watt variable LED illuminator, field and aperture diaphragms, filter slider
  • Daylight blue, green, and frosted filters
Specifications
Eyepiece WF 10X/23 mm