Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Monoclonal Antibody, anti-mouse PD-1, RMP1-14, 50 MG

Supplier:  Bio X Cell BP0146R050MG

Encompass_Preferred

The RMP1-14 monoclonal antibody reacts with mouse PD-1 (CD279), a cell surface receptor crucial for peripheral tolerance and autoimmune disease prevention. It inhibits T-cell activation upon ligand binding and blocks PD-L1-Ig and PD-L2-Ig binding to PD-1.

  • Clone: RMP1-14
  • Reactivities: Mouse
  • Isotype: Rat IgG2a, κ
  • Conjugation: Unconjugated
  • Formulation: PBS, pH 7.0
  • Production: Purified from animal-free facility by Protein G
  • Endotoxin: ≤0.5EU/mg
  • Aggregation: <5%
  • Purity: ≥95% (SDS-PAGE)
  • Sterility: 0.2 μm filtration
  • Molecular weight: 150 kDa
  • Storage: 4°C; do not freeze
  • Applications: In vivo PD-1/PD-L blocking, in vitro organoids

Catalog No. NC1385633


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.