Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Invisible Footprints: Seeing Carbon Dioxide and Understanding Climate Change Kit
SDPSCI_ED

Catalog No. S27557 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
20 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27557 20 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27557 Supplier Edvotek™ Supplier No. 930
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this experiment, students get to be in charge of their own personal atmosphere! Explores the global carbon cycle and climate change in a simple, positive, and intimate way. Ideal for a middle school classroom.

  • Completion Time: 1 Week (1-1.5 hours total lab time)
  • For 20 Students

Specifications

Grade 8 to Undergraduate
Age 14 to 20
Includes Instructions, Atmosphere Powder, CO2 e-powder, Color Change Solution, Mitigation Solution, Conical Tubes, Microcentrifuge Tubes, Transfer Pipettes, Carbon Footprint Questions, Carbon Handprint Actions
Requires Distilled Water, and 3 Beakers/Flasks/Cups
Time Range 1 Week (Total Lab Time: 75 min.)
Content And Storage Room Temperature
Class Size 20 Students
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.