Learn More
Electron Microscopy Sciences Invisible Glass 16 OZ
SDP
Catalog No. 5033444
Encompass_Preferred
Specially formulated, Invisible Glass quickly and easily removes dust, dirt, oil, grease, fingerprints, smoke film, haze and more from glass surfaces, such as windows, mirrors, computer screens, display cabinets, and photocopiers. Invisible Glass contains no water and soaps, surfactants or foams so it will leave no residue that will either streak or haze.
Catalog No. 50-334-44 Supplier Electron Microscopy Sciences Supplier No. 6081101
Specially formulated, Invisible Glass quickly and easily removes dust, dirt, oil, grease, fingerprints, smoke film, haze and more from glass surfaces, such as windows, mirrors, computer screens, display cabinets, and photocopiers. Invisible Glass contains no water and soaps, surfactants or foams so it will leave no residue that will either streak or haze.