Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BrightStar™-Plus Positively Charged Nylon Membrane, 30 cm x 3 m

Catalog No. AM10104
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM10104 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM10104 Supplier Invitrogen™ Supplier No. AM10104
  • $780.00
    $756.65 / Each of 1

    Save $23.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For use with radiolabeled and nonisotopic probes for northern and other blot applications

  • For consistent, reliable results with minimal background
  • One roll of 30cm x 3m is provided
  • This membrane exhibits exceptionally low nonspecific binding in nonisotopic detection applications when used with proprietary blocking and wash solutions

DNA and RNA Purification and Analysis, Dot Blotting, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Proteins, Expression, Isolation and Analysis, Southern Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Membrane Nylon Membrane
Quantity 1 roll
Shipping Condition Room Temperature
Product Type Nylon Membrane

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.