Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BioPrime™ Total Genomic Labeling Module

Catalog No. 18097012
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-097-012 30 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-097-012 Supplier Invitrogen™ Supplier No. 18097012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Supports a complete genomic DNA labeling system for use in array comparative genomic hybridization applications (aCGH) that allow for better call rates with precious samples

The BioPrime Total Genomic Labeling Module is part of the system that offers users the following advantages:

  • Higher signal to noise with Alexa Fluor™ 3 and 5 dyes
  • Less channel bias due to novel reaction formulation
  • Widest range of input material (50ng - 3μg)
  • Simplified workflow with master mix formulation
This complete, all-inclusive kit is optimized to work across the widest range of sample input material with no need for pre-amplification. Highly concentrated exo-Klenow fragment of DNA polymerase I in a component-limited reaction allows for consistent, robust DNA yields of approx. 8μg with as little as 50ng of genomic DNA input. An optimized dye-labeled nucleotide mix with new dNTP linkers, novel application-specific Alexa Fluor 3 and 5 dyes, and improved buffer chemistry reduce labeling variation and increase signal to noise on arrays. Excitation and emission spectra of Alexa Fluor 3 and 5 dyes are suited for conventional two color scanners with no need to change settings. New master mix formulation streamlines reaction set-ups, increasing consistency. Complete kits are batch tested to assure quality in performance from lot to lot.
Contents and Storage:
The BioPrime™ Total Genomic Labeling Module contains Alexa Fluor 3 and Alexa Fluor 5 2x reaction mix, Exo-Klenow fragment (40 U/μL), 5mM EDTA, TE Buffer, and control DNA (Salmon Sperm). Available in a direct labeling format.

Array CGH, ChIP-on-Chip, Chromatin Biology, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis, RNAi, Epigenetics and Non-Coding RNA Research

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Label or Dye Alexa Fluor 3, Alexa Fluor 5
Final Product Type Probes (Labeled DNA)
Labeling Target Genomic DNA
Includes Label or Dye Yes
Labeling Method Direct Labeling
Product Line BioPrime
Product Type Total Genomic Labeling Module
Quantity 30 Reactions
Why do you need more labeling product for a cDNA array than for a BAC array?

The reason for using more genomic DNA for the cDNA array is because the BAC arrays have much longer DNA fragments on the array than cDNA, therefore higher homology to the labeled DNA than the cDNA array. The fluorescence signals from the cDNA will be weaker than the BAC array, therefore, you have to start with higher amounts of starting materials.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.