Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Biotin-14-dCTP
GSA_VA

Catalog No. 19518018
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-518-018 50 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-518-018 Supplier Invitrogen™ Supplier No. 19518018
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Biotin-14-dCTP is supplied at 0.4mM in 100mM Tris-HCl (pH 7.5), 0.1mM EDTA.

Used for DNA biotinylation, an excellent, and sometimes superior alternative to conventional radioisotopic labeling

About Biotin-14-dCTP
Biotin-14-dCTP is a patented dCTP analog with biotin attached at the N4-position of the pyrimidine base by a 14-atom linker. This nucleotide can be efficiently incorporated into DNA by nick translation in the presence of dATP, dGTP, and dTTP, labeling DNA with terminal deoxynucleotide transferase, or random primer synthesis. The amount of material provided is sufficient for labeling up to 50μg of DNA.

Performance and quality testing
Purity is evaluated by reverse-phase HPLC.

  • Convenience and safety: probe preparation generates no radioactive waste
  • Stability: probes remain active longer than isotopically labeled probes
  • Versatility: biotinylated DNA can be used for immobilization and capture procedures via streptavidin binding

DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis

Order Info

Shipping Condition: Dry Ice

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeled Nucleotide
Concentration 0.4 mM
Content And Storage Biotin-14-dCTP is supplied at 0.4 mM in 100 mM Tris-HCl (pH 7.5), 0.1 mM EDTA. Store at -20°C.
Label or Dye Biotin & Other Hapten
Labeling Method Direct Labeling
Shipping Condition Dry Ice
Quantity 50 nmol
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.