Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BloodPrep™ DNA Purification Solution

Catalog No. 4342775
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-427-75 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-427-75 Supplier Invitrogen™ Supplier No. 4342775
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The BloodPrep™ DNA Purification Solution is a cell lysis and DNA purification solution for whole blood from any animal species, cultured cells, and buccal swabs for use with BloodPrep™ DNA chemistry.

The BloodPrep™ DNA Purification Solution is a cell lysis and DNA purification solution for whole blood from any animal species, cultured cells, and buccal swabs for use with BloodPrep™ DNA chemistry.

For Research Use Only. Not for use in diagnostics procedures.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Nucleic Acid Purification
Product Line BloodPrep
Quantity 1000 mL
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.