Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ cDNA Size Fractionation Columns

Catalog No. 18092015
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-092-015 3 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-092-015 Supplier Invitrogen™ Supplier No. 18092015
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Each 2mL plastic column contains 1mL of Sephacryl S-500 HR resin.

Offers efficient and convenient method for size-fractioning double-stranded cDNA (1)

  • Pre-packed, disposable columns efficiently remove small (<500 bp) cDNAs-including residual adapters, linkers, or other fragments that could interfere with ligation of cDNA to the vector
  • Columns size-fractionate cDNA >500 bp
  • Performance and Quality Testing: Wash time, drop volume during elution of fragments, and ability to clearly and cleanly separate mixture of dyes, including xylene cyanol and bromophenol blue, are evaluated

Cloning, cDNA Libraries and Library Construction

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Fractionation Column
Content And Storage Each 2 ml plastic column contains 1 ml of Sephacryl™ S-500 HR resin. Store at +4°C.
Shipping Condition Wet Ice
Format Pre-packed Column
For Use With (Application) cDNA Libraries & Library Construction
Quantity 3 Columns

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.