Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ M-270 Carboxylic Acid

Catalog No. p-4924201
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-305-D 2 mL
14-306-D 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-305-D Supplier Invitrogen™ Supplier No. 14305D
Only null left
Add to Cart
Edge
Add to Cart

These are hydrophilic Dynabeads™ (2.8 μm) with carboxylic acid groups. These surface groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups.

These surface groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups. For use with any sample type.

  • Immediate and gentle coupling at low temperature and neutral to low pH
  • Activation through carbodiimide is required
  • Alternative reactive groups can be introduced by using commercially available cross-linkers
  • Used for isolation of small components such as proteins and peptides
  • Immobilization of labile proteins/peptides and N-terminal coupling of peptides
  • Low non-specific binding of nucleic acids
  • High throughput compatible

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Diameter (Metric) 2.8 μm
Elution Volume Min 5 μL
High-throughput Compatibility High-throughput Compatible
Shipping Condition Room Temperature
Sample Type Any Sample Type
Surface Functionality Carboxylic Acid
Product Line DYNAL, Dynabeads
Quantity 2 mL
Type Carboxylic Acid Magnetic Bead
Format Beads in suspension
Purity or Quality Grade Research Grade
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.